IDEX® PEEK Tubing, 5 Ft Length

An excellent replacement for stainless steel tubing
  • Flexible PEEK tubing is biocompatible and chemically inert
  • Easily cut to desired lengths
  • Smooth internal surface decreases turbulence
  • Color-coded for easier identification
Applications: Chromatography
Characteristics: Unlike stainless steel, PEEK (polyetheretherketone) tubing is flexible and easy to cut. It provides excellent chemical resistance to both organic and inorganic solvents. Tubing withstands pressures to 5000 psi (3000 psi for model 02006-30). PEEK tubing has a very smooth internal surface, which causes less turbulence than metal tubing and improves resolution. Pack of 5-ft.
Certification: None
Maximum temperature: 212°F (100°C)
Sterilize: With autoclave, ethylene oxide, or gamma irradiation
$50.10 - $229.70CAD / Each

22 variations of this product are available.

See All Product Options Below
22 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Tubing ID (in) Tubing OD (in)
Availability Pricing
Idex 1531 Chromatography Tubing, Natural PEEK, 1/16" OD x 0.010" ID x 5 ft L; 1/EA 0200600 Mfr # 1531
COMPARE
Natural 1/100 1/16
$50.10CAD / Pkg of 1
Idex Chromatography Tubing, Natural PEEK, 1/8" OD x 0.062" ID x 5 ft L 0200601 Mfr # 1534
COMPARE
Natural 1/16 1/8
$103.73CAD / Each
Idex 1535 Chromatography Tubing, Red PEEK, 1/16" OD x 0.005" ID x 5 ft L; 1/EA 0200602 Mfr # 1535
COMPARE
Red 1/200 1/16
$71.16CAD / Each
Idex Chromatography Tubing, Yellow PEEK, 1/16" OD x 0.007" ID x 5 ft L 0200603 Mfr # 1536
COMPARE
Yellow 1/150 1/16
$50.10CAD / Each
Idex 1538 Chromatography Tubing, Natural PEEK, 1/16" OD x 0.040" ID x 5 ft L; 1/EA 0200604 Mfr # 1538
COMPARE
Natural 1/25 1/16
$50.10CAD / Each
Idex Chromatography Tubing, Natural PEEK, 1/8" OD x 0.080" ID x 5 ft L 0200605 Mfr # 1544
COMPARE
Natural 5/64 1/8
$102.70CAD / Each
Idex Chromatography Tubing, Natural PEEK, 1/16" OD x 25 µm ID x 5 ft L 0200606 Mfr # 1559
COMPARE
Natural 1/1000 1/16
$76.76CAD / Each
Idex 1560 Chromatography Tubing, Natural PEEK, 1/16" OD x 0.0025" ID x 5 ft L; 1/EA 0200607 Mfr # 1560
COMPARE
Natural 1/500 1/16
$71.16CAD / Each
Idex 1561 Chromatography Tubing, Black PEEK, 1/16" OD x 0.004" ID x 5 ft L; 1/EA 0200608 Mfr # 1561
COMPARE
Black 1/250 1/16
$71.16CAD / Each
Idex 1568 Chromatography Tubing, Natural PEEK, 1/32" OD x 0.015" ID x 5 ft L; 1/EA 0200609 Mfr # 1568
COMPARE
Natural 1/64 1/32
$85.01CAD / Each
Idex 1531B Chromatography Tubing, Blue PEEK, 1/16" OD x 0.010" ID x 5 ft L; 1/EA 0200610 Mfr # 1531B
COMPARE
Blue 1/100 1/16
$50.10CAD / Pkg of 1
Idex Chromatography Tubing, Orange PEEK, 1/32" OD x 0.020" ID x 5 ft L 0200611 Mfr # 1569
COMPARE
Orange 1/50 1/32
$85.01CAD / Each
Idex 1570 Chromatography Tubing, Natural PEEK, 360 µm OD x 50 µm ID x 5 ft L; 1/EA 0200612 Mfr # 1570
COMPARE
Natural 1/500 1/64
$86.93CAD / Each
Idex 1571 Chromatography Tubing, Red PEEK, 360 µm OD x 100 µm ID x 5 ft L; 1/EA 0200613 Mfr # 1571
COMPARE
Red 1/250 1/64
$85.01CAD / Each
Idex 1572 Chromatography Tubing, Yellow PEEK, 360 µm OD x 150 µm ID x 5 ft L; 1/EA 0200614 Mfr # 1572
COMPARE
Yellow 1/175 1/64
$85.01CAD / Each
Idex 1574 Chromatography Tubing, Natural PEEK, 360 µm OD x 25 µm ID x 5 ft L; 1/EA 0200615 Mfr # 1574
COMPARE
Natural 1/1000 1/64
$85.01CAD / Each
Idex 1576 Chromatography Tubing, Red PEEK, 1/32" OD x 0.005" ID x 5 ft L; 1/EA 0200616 Mfr # 1576
COMPARE
Red 1/200 1/32
$85.01CAD / Each
Idex Chromatography Tubing, Yellow PEEK, 1/32" OD x 0.007" ID x 5 ft L 0200617 Mfr # 1577
COMPARE
Yellow 1/150 1/32
$85.01CAD / Each
Idex 1581 Chromatography Tubing, Blue PEEK, 1/32" OD x 0.010" ID x 5 ft L; 1/EA 0200618 Mfr # 1581
COMPARE
Blue 1/100 1/32
$85.01CAD / Each
Idex Chromatography Tubing, Orange PEEK, 1/16" OD x 0.020" ID x 5 ft L 0200620 Mfr # 1532
COMPARE
Orange 1/50 1/16
$50.10CAD / Pkg of 1
Showing 1 - 20 of 22

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat