Idex® Low-Pressure Y Connector Assemblies

Combine, split, or intersect fluid streams throughout your system
  • Manufactured from inert polymers
Use Y connectors in a variety of low-pressure situations throughout your fluidic system. Y connectors create less turbulence and solvent outgassing than Tee connectors, improving analytical results. Handy mounting holes make it a snap to mount connectors. Y connector assemblies come complete with nuts and ferrules.
$59.67 - $72.93CAD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 2) (in) Thread size UNF(F) (port 1) (InchShort) Orifice Dimensions (mm)
Availability Pricing
Idex Low-Pressure Y Connector Assembly, Natural PEEK, 0.020" Bore, 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0201391 Mfr # P-512
COMPARE
1/16 1/4-28 Flat Bottom 0.5
$72.93CAD / Each
Idex Low-Pressure Y Connector Assembly, Natural PEEK, 0.060" Bore, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0201392 Mfr # P-514
COMPARE
1/8 1/4-28 Flat Bottom 1.5
$59.67CAD / Each
Idex Low-Pressure Y Connector Assembly, Natural PEEK, 0.020" Bore, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202331 Mfr # P-513
COMPARE
1/8 1/4-28 Flat Bottom 0.5
$60.70CAD / Each
Idex Low-Pressure Y Connector Assembly, Natural PEEK, 0.125" Bore, 3/16" OD Tubing, 5/16-24 Flat Bottom; 1/EA 0202334 Mfr # P-515
COMPARE
3/16 5/16-24 Flat Bottom 3.2
$60.70CAD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat