Idex® Low-Pressure Tee Bodies
Combine, split, or intersect fluid streams throughout your system
  • Manufactured from inert polymers
  • 1/4-28 flat-bottom ports
  • Provide your own fittings to suit your application
Use Tee connectors in a variety of low-pressure situations throughout your fluidic system. Handy mounting holes make it a snap to mount connectors.
$37.28 - $45.53CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Color Fitting Material Orifice Dimensions (mm)
Availability Pricing
Idex Low-Pressure Tee Body, Natural ETFE, 0.020" Bore, 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202348 Mfr # P-632-01
COMPARE
Natural ETFE 0.5
$43.02CAD / Each
Idex Low-Pressure Tee Body, Natural ETFE, 0.050" Bore, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202349 Mfr # P-633-01
COMPARE
Natural ETFE 1.25
$43.32CAD / Each
Idex Low-Pressure Tee Body, Natural PEEK, 0.020" Bore, 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202374 Mfr # P-712-01
COMPARE
Natural PEEK 0.5
$38.46CAD / Each
Idex Low-Pressure Tee Body, Red PEEK, 0.020" Bore, 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202375 Mfr # P-712-01R
COMPARE
Red PEEK 0.5
$45.53CAD / Each
Idex Low-Pressure Tee Body, Natural PEEK, 0.050" Bore, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202376 Mfr # P-713-01
COMPARE
Natural PEEK 1.25
$38.46CAD / Each
Idex Low-Pressure Tee Body, Natural PEEK, 0.040" Bore, 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202377 Mfr # P-714-01
COMPARE
Natural PEEK 1
$37.28CAD / Each