Idex® Inline Chromatography Check Valves
Spring-actuated sealing system add back-flow protection to your system
  • Manage directional flow of fluid streams
  • Excellent chemical resistance
The 1/4-28 flat bottom valves can be used with any 1/4-28 flangeless, Super Flangeless™, and VacuTight™ fittings. The 10-32 coned bottom valve can be used with 10-32 polymer fingertight or SealTight™ fittings. Select from inlet or outlet check valves.
$127.45 - $206.86CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Breaking Pressure (PSI) Inlet Port Orifice (in)
Availability Pricing
Idex Inline Check Valve, Inlet, 3 psi, 0.02" ID, 1/4-28 (F) to 1/4-28 (M) Flat Bottom; 1/EA 0201334 Mfr # CV-3315
COMPARE
3 1/4-28 (F) flat bottom 0.02
$206.86CAD / Each
Idex Inline Check Valve, Outlet, 3 psi, 0.02" ID, 1/4-28 (M) to 1/4-28 (F) Flat Bottom; 1/EA 0201335 Mfr # CV-3316
COMPARE
3 1/4-28 (M) flat bottom 0.02
$206.86CAD / Each
Idex Inline Check Valve, Inlet, 15 psi, 0.02" ID, 1/4-28 (F) to 1/4-28 (M) Flat Bottom; 1/EA 0201588 Mfr # CV-3301
COMPARE
15 1/4-28 (F) flat bottom 0.02
$127.45CAD / Each
Idex Inline Check Valve, Outlet, 15 psi, 0.02" ID, 1/4-28 (M) to 1/4-28 (F) Flat Bottom; 1/EA 0201589 Mfr # CV-3302
COMPARE
15 1/4-28 (M) flat bottom 0.02
$127.45CAD / Each
Idex Inline Check Valve, Outlet, 3 psi, 0.04" ID, 1/4-28 (M) to 1/4-28 (F) Flat Bottom; 1/EA 0201590 Mfr # CV-3310
COMPARE
3 1/4-28 (F) flat bottom 0.04
$204.80CAD / Each
Idex Inline Check Valve, Outlet, 15 psi, 0.02" ID, 10-32 (M) to 10-32 (F) Coned; 1/EA 0201596 Mfr # CV-3345
COMPARE
15 10-32 (M) coned bottom 0.02
$201.12CAD / Each