Idex® Chromatography Tubing Cutters

Easily make clean cuts with tubing cutters
  • Cutters are available for different tubing sizes and materials
Choose from tubing cutters for a wide range of tubing sizes, suitable for a wide range of applications.
$48.92 - $956.23CAD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
For Tubing OD (in) Max Tubing OD (in) Description
Availability Pricing
Idex Polymer Tubing Cutter For 1/16" to 1/8" OD Tubing; 1/EA 0201329 Mfr # A-327
COMPARE
1/16 1/8 Polymer Tubing Cutter For 1/16" to 1/8" OD Tubing; 1/EA
$48.92CAD / Each
Idex Capillary Polymer Chromatography Tubing Cutter For 360 µm to 1/32" OD tubing; 1/EA 0201330 Mfr # A-350
COMPARE
1/64 1/32 Capillary Polymer Chromatography Tubing Cutter For 360 µm to 1/32" OD tubing; 1/EA
$266.39CAD / Each
Idex Polymer Tubing Cutter For 3/16" to 5/16" OD Tubing; 1/EA 0201687 Mfr # A-329
COMPARE
3/16 5/16 Idex A-329 Polymer Tubing Cutter For 3/16" to 5/16" OD Tubing; 1/EA
$49.65CAD / Each
Idex Fused Silica Tubing Cutter For 350 to 780 µm OD Tubing; 1/EA 0201688 Mfr # FS-315
COMPARE
1/75 1/32 Idex FS-315 Fused Silica Tubing Cutter For 350 to 780 µm OD Tubing; 1/EA
$956.23CAD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat