Idex® Chromatography Inline Cartridge Check Valves
Add back-flow protection to your system
  • Low breaking pressures
  • Less than 150 µL internal volume
  • Excellent chemical resistance
Use these inline check valves for applications requiring operator control or low frequency of use. Valves are available with or without fittings.
$175.19 - $188.59CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Breaking Pressure (PSI) Inlet Port Outlet Port
Availability Pricing
Idex Inline Cartridge Check Valve Assembly, 1.5 psi, 1/16" OD Tubing; 1/EA 0201333 Mfr # CV-3000
COMPARE
1.5 1/16" tubing OD (1/4-28 flat bottom) 1/16" tubing OD (1/4-28 flat bottom)
$188.59CAD / Each
Idex Inline Cartridge Check Valve Assembly, 1.5 psi, No Fittings; 1/EA 0201584 Mfr # CV-3000NF
COMPARE
1.5 1/4-28 (F) flat bottom 1/4-28 (F) flat bottom
$175.19CAD / Each
Idex Inline Cartridge Check Valve Assembly, 3 psi, 1/8" OD Tubing; 1/EA 0201586 Mfr # CV-3010
COMPARE
3 1/8" tubing OD (1/4-28 flat bottom) 1/8" tubing OD (1/4-28 flat bottom)
$183.00CAD / Each