Idex® Actuated Rotary Shear Selection Valves

Automated valves for rapid, consistent flow distribution
  • Control either manually or remotely, with software or instrument
  • Operate at higher torque levels or very small flow paths
Automated rotary shear valves are designed for applications that require fast, consistent fluid control and delivery. Use selection valves to distribute a variety of system liquids by a common port. Connect valves to your PC or instrument via USB, BCD, serial port, or contact closure.
$3,368.17 - $3,598.02CAD / Each

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Description Flow Pattern Max Pressure (PSI)
Availability Pricing
Idex Actuated Selection Valve, 6-Position, 7-Port, 125 psi, RPC-7; 1/EA 0201609 Mfr # MXX777-605
COMPARE
Actuated Selection Valve, 6-Position, 7-Port, 125 psi, RPC-7; 1/EA 6-position, 7-port 125
$3,368.17CAD / Each
Idex Actuated Selection Valve, 10-Position, 11-Port, 125 psi, RPC-7; 1/EA 0201611 Mfr # MXX778-605
COMPARE
Actuated Selection Valve, 10-Position, 11-Port, 125 psi, RPC-7; 1/EA 10-position, 11-port 125
$3,598.02CAD / Each
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat