Ice-Pak Soft Cold Packs
Ideal for shipping perishable or heat-sensitive materials – concentrated cold lasts longer than ice
The refrigerant is designed to protect your product from freezing or over-heating even in extreme conditions. Gel filling is nontoxic, nonhazardous and biodegradable; won't contaminate samples. Reusable packs freeze solid in 6 to 10 hours and remain at constant temperature for up to 72 hours. Place packs in the freezer for 24 hours before use.
$113.01CAD / Pkg of 12
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Inside Width (in) Min Temperature (° C) Shipping Weight (oz)
Availability Pricing
Cryopak Ice-Pak Reusable Gel-Filled Ice Packs; 0 degrees C, 9" x 6-1/2" x 1" cs/12 0634605 Mfr # 81111
COMPARE
0 26.5
$113.01CAD / Pkg of 12