Cryopak Ice-Pak Reusable Gel-Filled Ice Packs; 0 degrees C, 9" x 6-1/2" x 1" cs/12

Representative image only.

Cryopak Ice-Pak Reusable Gel-Filled Ice Packs; 0 degrees C, 9" x 6-1/2" x 1" cs/12

Cryopak | Mfr: 81111| Item: 0634605

Ideal for shipping perishable or heat-sensitive materials – concentrated cold lasts longer than ice

$113.01CAD / Pkg of 12

$113.01CAD / Pkg of 12
Product Description

Key Features

  • 1 year warranty

More About this Item

The refrigerant is designed to protect your product from freezing or over-heating even in extreme conditions. Gel filling is nontoxic, nonhazardous and biodegradable; won't contaminate samples. Reusable packs freeze solid in 6 to 10 hours and remain at constant temperature for up to 72 hours. Place packs in the freezer for 24 hours before use.

Gel changes phase (freeze to thaw) at 0° C (32°F)

1 Year Warranty
Specifications
  • Min Temperature (° C)0
  • Min Temperature (° F)32
  • Outside Width (in)6 1/2
  • Outside Length (in)9
  • Outside Height (in)1
  • Outside Width (cm)16.5
  • Outside Length (cm)22.9
  • Outside Height (cm)2.5
  • Shipping Weight (oz)26.5
  • Shipping Weight (g)751
  • DescriptionReusable Gel-Filled Ice Packs; 0 degrees C, 9" x 6-1/2" x 1" cs/12
  • Warranty1 year
Reviews
Q&A