Honeywell DR Graphic Recorder
Go paperless!
  • Identical panel cutout to Honeywell DR4300 and DR4500 aids the transition away from chart recording
  • Chart-paper-like display is familiar and easy to read
  • Touch screen, data storage, and communications features make programming simple and expand capabilities
This paperless recorder lets you upgrade from traditional chart paper recording to paperless recording without requiring a new panel build if you have the Honeywell DR4300 or DR4500 recorder currently. The display mimics a circular chart, so it is visually similar, but without the inconvenience and potential downtime associated with pens and chart paper.
Use the supplied configuration credits to customize your recorder. Each model is programmed with ten credits to use in any way. All functions are programmed in the firmware, but must be unlocked using the credits. You can mix and match and reprogram at any time. Available options include maintenance, printing, extra digital pens, configuration lockout, remote viewing, reports, e-mail, secure communications, custom screens, maths, and much more.
Choose your software:
TrendManager Pro software offers all of the functions of TrendViewer and adds full recorder configuration and data export capabilities.
TrendServer Pro software builds on the features of TrendManager Pro by adding network connectivity with FTP and web browser access and real-time data acquisition.
Many more input/output, power, memory, and credit configurations are available through special order.
$14,278.62 - $16,012.80CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Honeywell DR Graphic Recorder with TrendManager Pro Software RK-49031-05 Mfr # TVDRG2-040-12-0-010-00000P-000
COMPARE
Graphic Recorder with TrendManager Pro Software
$14,278.62CAD / Each
Honeywell DR Graphic Recorder with TrendServer Pro Software RK-49031-07 Mfr # TVDRG2-040-12-0-010-00000S-000
COMPARE
Graphic Recorder with TrendServer Pro Software
$16,012.80CAD / Each