HF® scientific MTOL+™ Online Process Turbidimeter
Take advantage of advanced data storage capabilities while maximizing accuracy and cleanliness
  • Ultrasonic autoclean system keeps optical chamber clean
  • Customize data logging to suit your needs
  • Bidirectional RS-485 and Modbus RTU
  • Two programmable alarms rated at 120 to 240 VAC, 2 A
  • 4 to 20 mA output
Completely electronically redesigned, the MTOL+ online process turbidimeter employs a 32 bit ARM Core Processor. You're now able to log/store a year's worth of measurement/calibration data, install software updates via a USB flash drive and download data. The unit features simultaneous 4 to 20 mA and Modbus signal outputs, and variable user-selectable logging interval from 1 to 60 minutes. Arrives ready to accommodate a range of 0 to 100 NTU; calibration files enabling 0 to 10 NTU and 0 to 1000 NTU are included on a USB key.
$6,829.16 - $7,069.33CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
HF Scientific MTOL+ Online Turbidimeter with Autoclean and Datalogger, White Light; 0 to 100 NTU RK-99544-30 Mfr # 28053
COMPARE
MTOL+ Online Turbidimeter with Autoclean and Datalogger, White Light; 0 to 100 NTU
$6,829.16CAD / Each
HF Scientific MTOL+ Online Turbidimeter with Autoclean and Datalogger, IR; 0 to 100 NTU RK-99544-35 Mfr # 28052
COMPARE
MTOL+ Online Turbidimeter with Autoclean and Datalogger, IR; 0 to 100 NTU
$7,069.33CAD / Each