Hemco Universal Ducted Fume Hoods
Designed for light- to medium-duty fume removal
  • Various sizes which is ideal for tight spaces in the laboratory
  • Fire-retardant fiberglass construction with shatterproof sash adds user safety
  • Dual walled design creates a quieter area around the hood
  • One-piece interior with coved corners for easy cleaning
  • Shipped completely assembled which assists in a quicker setup
The universal fume hood is available in 24", 30", 36", and 48" widths which can cover all space limitations in the laboratory. The fully assembled model comes with a vapor-proof light switch and a 3-wire cord. There is also an option that includes an exhaust blower and another that adds an explosion-proof light. These need some additional assembly by the user. The explosion-proof light model is UL approved Class 1, Group D, Class II, Group E,F, and G.
$9,252.89 - $13,680.43CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Hood Width (in) Power (VAC) Description
Availability Pricing
HEMCO Universal Fume Hood w/ Vapor Proof Light, 24" W; 115 VAC 3370600 Mfr # 92021
COMPARE
24 115 Universal Fume Hood w/ Vapor Proof Light
$9,378.13CAD / Each
HEMCO Universal Fume Hood w/ Vapor Proof Light, 30" W; 115 VAC 3370601 Mfr # 93021
COMPARE
30 115 Universal Fume Hood w/ Vapor Proof Light
$9,252.89CAD / Each
HEMCO Universal Fume Hood w/ Vapor Proof Light, 36" W; 115 VAC 3370602 Mfr # 90321
COMPARE
36 115 Universal Fume Hood w/ Vapor Proof Light
$11,124.09CAD / Each
HEMCO Universal Fume Hood w/ Blower, 30" W; 115 VAC 3370609 Mfr # 93022
COMPARE
30 115 Universal Fume Hood w/ Blower
$10,763.11CAD / Each
HEMCO Universal Fume Hood w/ Blower, 36" W; 115 VAC 3370610 Mfr # 90322
COMPARE
36 115 Universal Fume Hood w/ Blower
$13,680.43CAD / Each