Hamilton® SoftGrip™ Adjustable-Volume Pipettors
Contoured, ergonomic design provides comfortable, fatigue-free operation
  • Less actuation force minimizes thumb and hand strain
  • Color coding allows easy volume identification
  • Autoclave entire pipettor for total decontamination and sterilization
SoftGrip™ pipettors assure you of quality, comfort, and precision in manual pipetting. The isolated volume adjustment makes it so there's no need for a locking mechanism to prevent accidental volume adjustment. Our new generation of pipettes has a dual indicator that improves the volume adjustment accuracy by removing the parallax errors. The slim ejection sleeve is made from a high strength, chemically resistant polymer. The slim design allows the pipette to reach to the bottom of narrow tubes without the need for extended length tips. SoftGrip pipettes have a universal calibration key that reduces the need for costly calibration services. Calibration requires only minutes to perform and eliminates the downtime and cost of alternative calibration methods. The SoftGrip tip nozzle ensures a proper fit with universal pipette tips. Unique color coding and labeling clearly indicates the working ranges of the adjustable volume pipettes.
$474.43 - $512.00CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Increments (µL) Max Volume (µL) Min Volume (µL)
Availability Pricing
Hamilton 55019-32 Adjustable-Volume Pipettor, 1-10 µL; Each 2570402 Mfr # 55019-32
COMPARE
0.02 10 1
$512.00CAD / Each
Hamilton SoftGrip™ Adjustable-Volume Pipettor, Violet, 10-100 µL 2570406 Mfr # 55019-36
COMPARE
0.2 100 10
$474.43CAD / Each
Hamilton SoftGrip™ Adjustable-Volume Pipettor, Brick Red, 30-300 µL 2570408 Mfr # 55019-38
COMPARE
0.2 300 30
$474.43CAD / Each
Hamilton SoftGrip™ Adjustable-Volume Pipettor, Sky Blue, 100-1000 µL 2570410 Mfr # 55019-40
COMPARE
2 1000 100
$474.43CAD / Each