GLI 1000 Ω Pt RTD Contacting Conductivity Cells

Contacting conductivity measurements from ultra-pure to high conductivity measurements
  • Suitable for GLI Contacting Conductivity Controller and Transmitter
Nonmetallic Ryton® PPS cells 19065-55 and -57 have graphite electrode with 3⁄4" NPT(M) threads for mounting onto a 3⁄4" pipe tee or immersion applications. Sensor includes built-in 1000 Ω Pt RTD and 20-ft (6-m) cable.
PVDF cells 19065-60 through -68 have titanium electrodes with either 1⁄2" or 3⁄4" NPT(M) compression fitting for mounting onto a pipe tee with up to four inches of insertion depth. Reversing the compression fitting enables the sensor to be fastened onto the end of an immersion pipe. Cells include built-in 1000 Ω Pt RTD and 20-ft (6-m) cable.
$1,155.14 - $1,396.77CAD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Hach 3433E8A Contacting Conductivity Cell, 1000 ohm Pt RTD, graphite, K=10 1906557 Mfr # 3433E8A
COMPARE
Contacting Conductivity Cell, 1000 ohm Pt RTD, graphite, K=10
$1,155.14CAD / Each
Hach 3422A1A Contacting Conductivity Cell, 1000 ohm Pt RTD, titanium, K=0.05 1906560 Mfr # 3422A1A
COMPARE
Contacting Conductivity Cell, 1000 ohm Pt RTD, titanium, K=0.05
$1,396.77CAD / Each
Hach 3422B3A Contacting Conductivity Cell, 1000 ohm Pt RTD, titanium, K=0.5 1906562 Mfr # 3422B3A
COMPARE
Contacting Conductivity Cell, 1000 ohm Pt RTD, titanium, K=0.5
$1,396.77CAD / Each
Hach 3422C3A Contacting Conductivity Cell, 1000 ohm Pt RTD, titanium, K=1.0 1906564 Mfr # 3422C3A
COMPARE
Contacting Conductivity Cell, 1000 ohm Pt RTD, titanium, K=1.0
$1,396.77CAD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat