Glas-Col Flexible Fabric Separatory Funnel Heating Mantles
Get better heating with conforming flexible fabric
Heating Mantles feature insulated glass fabric that conforms to unusual vessel shapes. Separatory Funnel Mantles allow the extraction of solutions while hot, preventing solidification
of viscous material in cold stopcocks. Design prevents separatory funnel from cracking under extreme thermal strain.Mantles require a controlled electrical power source to prevent overheating.
$1,164.71 - $1,404.88CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Max temperature (° F) Power (VAC)
Availability Pricing
Glas-Col 100AO852 Separatory Funnel Mantle 500 ml Funnel, 180 Watts, 115 V; Squibb 3622782 Mfr # 100AO852
COMPARE
500 842 115
$1,164.71CAD / Each
Glas-Col 100AO854 Separatory Funnel Mantle 1000 ml Funnel, 190 Watts, 115 V; Squibb 3622784 Mfr # 100AO854
COMPARE
1000 842 115
$1,404.88CAD / Each