Glas-Col Flexible Fabric Buchner Funnel Heating Mantles
Get better heating with conforming flexible fabric
Heating Mantles feature insulated glass fabric that conforms to unusual vessel shapes. Büchner Funnel Mantles provide gentle heat for solutions that must be filtered while warm. Mantles require a controlled electrical power source to prevent overheating.
$883.89 - $1,474.86CAD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Coors Funnel Number Power (VAC)
Availability Pricing
Glas-Col 100A0817 Buchner Funnel Mantle for Coors Funnel No. 60245, 100 Watts, 230 V 3622755 Mfr # 100A O817
COMPARE
60245 230
$1,215.69CAD / Each
Glas-Col 100AO808 Buchner Funnel Mantle for Coors Funnel No. 60240, 50 Watts, 115 V 3622764 Mfr # 100AO808
COMPARE
60240 115
$883.89CAD / Each
Glas-Col 100AO812 Buchner Funnel Mantle for Coors Funnel No. 60243, 70 Watts, 115 V 3622768 Mfr # 100AO812
COMPARE
60243 115
$1,217.02CAD / Each
Glas-Col 100AO814 Buchner Funnel Mantle for Coors Funnel No. 60244, 70 Watts, 115 V 3622770 Mfr # 100AO814
COMPARE
60244 115
$1,129.35CAD / Each
Glas-Col 100AO818 Buchner Funnel Mantle for Coors Funnel No. 60246, 220 Watts, 115 V 3622774 Mfr # 100AO818
COMPARE
60246 115
$1,220.11CAD / Each
Glas-Col 100AO820 Buchner Funnel Mantle for Coors Funnel No. 60247, 300 Watts, 115 V 3622776 Mfr # 100AO820
COMPARE
60247 115
$1,440.53CAD / Each
Glas-Col 100AO822 Buchner Funnel Mantle for Coors Funnel No. 60248, 350 Watts, 115 V 3622778 Mfr # 100AO822
COMPARE
60248 115
$1,474.86CAD / Each
Showing 1- 7 of 7