Glas-Col Flexible Fabric Beaker Heating Mantles
Get better heating with conforming flexible fabric
Heating Mantles feature insulated glass fabric that conforms to unusual vessel shapes. Griffin Beaker Mantles concentrate the heat at the bottom of the mantle for evensmall volume heating
applications. Mantles require a controlled electrical power source to prevent overheating.
$631.79 - $1,017.52CAD / Each
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flask Size (mL) Power (VAC) Power (watts)
Availability Pricing
Glas-Col 100AO602 Heating Mantle for 100 ml Beaker, 100 Watts, 115 V 3622702 Mfr # 100AO602
COMPARE
100 115 100
$631.79CAD / Each
Glas-Col 100AO604 Heating Mantle for 150 ml Beaker, 100 Watts, 115 V 3622704 Mfr # 100AO604
COMPARE
150 115 100
$700.60CAD / Each
Glas-Col 100AO606 Heating Mantle for 250 ml Beaker, 140 Watts, 115 V 3622706 Mfr # 100AO606
COMPARE
250 115 140
$653.45CAD / Each
Glas-Col 100AO609 Heating Mantle for 400 ml Beaker, 240 Watts, 230 V 3622707 Mfr # 100AO609
COMPARE
400 230 240
$700.01CAD / Each
Glas-Col 100AO608 Heating Mantle for 400 ml Beaker, 240 Watts, 115 V 3622708 Mfr # 100AO608
COMPARE
400 115 240
$667.45CAD / Each
Glas-Col 100AO611 Heating Mantle for 600 ml Beaker, 325 Watts, 230 V 3622709 Mfr # 100AO611
COMPARE
600 230 325
$823.18CAD / Each
Glas-Col 100AO610 Heating Mantle for 600 ml Beaker, 325 Watts, 115 V 3622710 Mfr # 100AO610
COMPARE
600 115 325
$745.09CAD / Each
Glas-Col 100AO612 Heating Mantle for 800 ml Beaker, 350 Watts, 115 V 3622712 Mfr # 100AO612
COMPARE
800 115 350
$859.58CAD / Each
Glas-Col 100AO615 Heating Mantle for 1000 ml Beaker, 430 Watts, 230 V 3622713 Mfr # 100AO615
COMPARE
1000 230 430
$951.81CAD / Each
Glas-Col 100AO614 Heating Mantle for 1000 ml Beaker, 430 Watts, 115 V 3622714 Mfr # 100AO614
COMPARE
1000 115 430
$979.80CAD / Each
Glas-Col 100AO616 Heating Mantle for 2000 ml Beaker, 550 Watts, 115 V 3622716 Mfr # 100AO616
COMPARE
2000 115 550
$937.81CAD / Each
Glas-Col 100AO618 Heating Mantle for 3000 ml Beaker, 630 Watts, 115 V 3622718 Mfr # 100AO618
COMPARE
3000 115 630
$1,017.52CAD / Each
Glas-Col 100AO620 Heating Mantle for 4000 ml Beaker, 710 Watts, 115 V 3622720 Mfr # 100AO620
COMPARE
4000 115 710
$999.40CAD / Each
Showing 1- 13 of 13