Glas-Col Aluminum Beaker Heating Mantles
Get uniform temperature control for low-volume samples
  • Rigid housing provides strength and durability while supporting the vessel weight
  • Fabric interior minimizes thermal shock and damage to glassware
  • Maximum temperature of 842°F (450°C)
Griffin beaker heating mantles have a resilient and pliable interior to cradle fragile beakers. These heating mantles have a resilient and pliable interior to safely cradle your fragile glass beakers.
Heat is concentrated at the bottom allowing small liquid volumes to be heated. Maximum heating temperature is 842°>F (450°>C). Mantles require a controlled electrical power source to prevent overheating.
$745.68 - $1,062.17CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Beaker Size (mL) Power (VAC)
Availability Pricing
Glas-Col 100B TM610 Heating Mantle, 600 ml, 325 Watts, 115 V 3623500 Mfr # 100B TM610
COMPARE
600 115
$745.68CAD / Each
Glas-Col 100B TM611 Heating Mantle, 600 ml, 325 Watts, 230 V 3623506 Mfr # 100B TM611CE
COMPARE
600 230
$966.99CAD / Each
Glas-Col 100B TM614 Heating Mantle, 1000 ml, 430 Watts, 115 V 3623510 Mfr # 100B TM614
COMPARE
1000 115
$789.88CAD / Each
Glas-Col 100B TM615 Beaker Heating Mantle, 1000 ml, 430 Watts, 230 V 3623516 Mfr # 100B TM615CE
COMPARE
1000 230
$1,062.17CAD / Each