GK Heller Dual-Shaft Mixer with Remote Speed Controller
Two shafts provide both low- and high-speed mixing Controller offers stepless speed control and reversible rotation
This dual-shaft mixer maintains constant speed regardless of changes in viscosity or line voltage. Use the direct-drive shaft for high-speed mixing of low-viscosity fluids; switch to the 12:1 reduction gear shaft for low-speed mixing of high-viscosity fluids.
The totally enclosed, 1/40-hp, permanent magnet motor is equipped with permanently lubricated bearings. Built-in drive current filters eliminate pulsing, reduce motor heating, and increase service life. Motor is connected to the remote controller via a 6-ft insulated cable, allowing you to place the controller away from the mixer—safe from splashes and spills. Solid-state remote controller features a forward/off/reverse switch, a stepless speed control dial, and “power on” indicator light.
What's included:
  • two 304 SS paddle assemblies (1-1/2" dia and 2-3/4" dia impellers on 8-3/4"L shafts)
  • keyless chuck [accepts up to 3/8" (10 mm) dia shafts]
  • dual-rod clamp
  • 8"L x 11/16" dia mounting rod
  • 6-ft, three-wire power cord
$2,932.05 - $3,272.40CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Torque (in-oz) Motor hp Power (VAC)
Availability Pricing
GK Heller GT31 Dual-shaft mixer with remote speed controller, 115 VAC 0466100 Mfr # 099D GT31
COMPARE
57.6 1/40 115
$2,932.05CAD / Each
GK Heller GT331 Dual-shaft mixer with remote speed controller, 230 VAC 0466102 Mfr # 099D GT331
COMPARE
57.6 1/40 230
$3,272.40CAD / Each