Fowler Euro-Cal IV Electronic Calipers

Get direct inch/metric/fraction conversion
  • Absolute/incremental measurement
  • Reads in inch, metric and fractions
  • Resolution: 0.0005"/0.01 mm
  • 4-way measuring
  • Stainless steel frame
  • IP54 level resistant to coolant and fluid
$448.28 - $690.65CAD / Each

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max range (in)
Availability Pricing
Fowler 54-100-330 Euro-Cal IV Electronic Caliper, 6"/150 mm 0992318 Mfr # 54-100-330-1
COMPARE
6
$448.28CAD / Each
Fowler 54-100-332-0 Euro-Cal IV Electronic Caliper, 12"/300 mm 0992321 Mfr # 54-100-332-0
COMPARE
12
$690.65CAD / Each
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat