Fluke Ti Series Professional Thermal Imagers
Quick transfer of images directly to PC, iPhone® or iPad® with Wi-Fi connectivity
  • 3.5" Color touch screen with selectable on-screen viewing modes
  • 5 megapixel digital visible light camera with lamp
  • Thermal fusion to blend IR with visible light images
  • Laser pointer and LaserSharp Auto Focus—uses a laser to automatically and accurately focus the camera
  • Adjustable emissivity
  • Field-changeable, rechargeable batteries with charge level indicators
  • User-friendly touch screen controls
  • GPS positioning
  • Streaming video
  • High temperature, low temperature and isotherm alarms
  • Wi-Fi app connectivity
  • Voice & IR-PhotoNotes™ annotation
These thermal imagers are fully radiometric. They measure and store temperatures at every point in the image. Fusion® technology provides enhanced problem detection and analysis capabilities through the blending of visual and infrared images. The built-in laser pointer also helps associate the physical target with the hot spot on the thermal image, indicating where a problem exists. Designed to withstand a 6.5 ft (2 m) drop.
These imagers feature both Wi-Fi and Bluetooth® connections. To activate the Wi-Fi function, simply download the Fluke Viewer app to your Apple® mobile device and link to your thermal imager. Then transfer images to your mobile device for further analysis, creating reports, and e-mailing results. You can also link the camera to your existing Bluetooth® headset (not included), allowing for record notes to be added to an image.
Fast and convenient menu navigation using either rugged touch screen or optimized tactile buttons. All data is stored on an SD memory card (included). Additional imager features include large built-in camera lens cover and SmartView® software. SmartView® software is a powerful modular suite of software tools for viewing, annotating, editing, and analyzing infrared images.
Fluke Connect with ShareLive video - Make the best decisions faster than ever before by viewing all temperature, mechanical, electrical and vibration measurements for each equipment asset in one place. Get started saving time and increasing your productivity. Fluke Connect with ShareLive video call is the only wireless measurement system that lets you stay in contact with your entire team without leaving the field. The Fluke Connect mobile app is available for Android and iOS and works with over 20 different Fluke products.
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Detector Size Max Temperature (° F) Pixels / Thermal Resolution
Availability Pricing
Fluke Ti300+ 60 Hz Professional Thermal Imager, 76,800 Pixels in Infrared Mode 2003757 Mfr # FLK-Ti300+ 60HZ
COMPARE
30 x 240 1202 76800
$15,470.58CAD / Each
Fluke Ti480 PRO 60 Hz Professional Thermal Imager, 307,200 Pixels in Infrared Mode, 614,400 Pixels in SuperResolution Mode 3974947 Mfr # FLK-Ti480 PRO 60HZ
COMPARE
640 x 480 (1280 x 960 with SuperResolution) 1832 307,200 (1,228,800 with SuperResolution)
$22,100.84CAD / Each
Fluke Ti401 PRO 60 Hz Professional Thermal Imager; 307,200 Pixels in Infrared Mode 3974957 Mfr # FLK-TI401-PRO 60HZ
COMPARE
640 x 480 1202 307,200
$18,564.70CAD / Each