Fluke® Solmetric PVA-1500 Series PV Analyzer I-V Curve Tracers
Accurately assess the health and performance of solar photovoltaic (PV) modules and arrays
  • Uses I-V (current versus voltage) curve tracing for quick and detailed performance data
  • Test up to 1500 V and up to 30 A*
  • Sweep-to-sweep delay of 9 seconds to measure 3.5 MW within an hour
  • Intuitive user interface for easy navigation and real-time analysis
  • SolSensor™ PV reference sensor measures irradiance and temperature
  • Take measurements even in hot environments
  • Wireless connection helps with quick setup and mobility
Enhance the output and prolong the service life of solar modules and arrays by accurately assessing the health and performance with the PVA-1500 PV analyzer I-V curve tracers. These high-precision analyzers use I-V curve tracing, the most complete solar module performance measurement, to quickly obtain detailed performance data of PV modules. Use the I-V curve tracers to pinpoint problems early to maximize solar energy production while minimizing downtime.
Clamp the included wireless SolSensor™ PV reference sensor to the module frame to measure irradiance and module temperature. With the angular response of each unit calibrated for rotation and elevation, the temperature-compensated sensor offers excellent accuracy over a broad range of technologies, sky conditions, and sun angles.
A portable design and an intuitive interface make it easy to use the analyzer in the field. The PV analyzer and the SolSensor sensor communicate wirelessly with a tablet or a laptop (Windows® only) via Wi-Fi. Download the Data Analysis Tool software to display the data in multiple, easy-to-read formats and a statistical overview of the entire array. The software looks at all the collected data for comparison to see where there are differences, identifying the issue for further troubleshooting. Easily create and send customized reports.
$20,447.69 - $32,747.55CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
DC Max (amps) DC Max (Volts) Description
Availability Pricing
Fluke Solmetric PVA-1500T2 PV Analyzer I-V Curve Tracer 1778503 Mfr # FLUKE-PVA-1500T2
COMPARE
30 1500 Solmetric PVA-1500T2 PV Analyzer I-V Curve Tracer
$20,447.69CAD / Each
Fluke Solmetric PVA-1500HE2 High-Efficiency PV Analyzer I-V Curve Tracer 1778504 Mfr # FLUKE-PVA-1500HE2
COMPARE
30 1500 Solmetric PVA-1500HE2 High-Efficiency PV Analyzer I-V Curve Tracer
$25,759.26CAD / Each
Fluke Solmetric PVA-1500/FPC High-Efficiency PV Analyzer I-V Curve Tracer with 1 Yr Premium Care 1778511 Mfr # FLUKE-PVA-1500/FPC
COMPARE
30 1500 Solmetric PVA-1500/FPC High-Efficiency PV Analyzer I-V Curve Tracer with 1 Yr Premium Care
$28,353.90CAD / Each
Fluke Solmetric PVA-1500/FPC3 High-Efficiency PV Analyzer I-V Curve Tracer with 3 Yr Premium Care 1778512 Mfr # FLUKE-PVA-1500/FPC3
COMPARE
30 1500 Solmetric PVA-1500/FPC3 High-Efficiency PV Analyzer I-V Curve Tracer with 3 Yr Premium Care
$32,747.55CAD / Each