Fluke Solmetric PVA-1500/FPC3 High-Efficiency PV Analyzer I-V Curve Tracer with 3 Yr Premium Care

Representative image only.

Fluke Solmetric PVA-1500/FPC3 High-Efficiency PV Analyzer I-V Curve Tracer with 3 Yr Premium Care

Fluke | Mfr: FLUKE-PVA-1500/FPC3| Item: 1778512

Accurately assess the health and performance of solar photovoltaic (PV) modules and arrays

$32,747.55CAD / Each

$32,747.55CAD / Each
Product Description

Includes

SolSensor sensor, clamp, MC4 wrenches, test lead sets (banana to alligator clip and banana to MC4), two type K thermocouple sensors, 50 adhesive discs for thermocouple attachment, sensor cleaning supplies, charging supplies, soft case, and hard carrying case.

Key Features

  • Uses I-V (current versus voltage) curve tracing for quick and detailed performance data
  • Test up to 1500 V and up to 30 A*
  • Sweep-to-sweep delay of 9 seconds to measure 3.5 MW within an hour
  • Intuitive user interface for easy navigation and real-time analysis
  • SolSensor™ PV reference sensor measures irradiance and temperature
  • Take measurements even in hot environments
  • Wireless connection helps with quick setup and mobility
  • High-efficiency I-V curve tracer
  • 1 year standard warranty
  • Three years of premium care provides coverage above and beyond the original warranty

More About this Item

Enhance the output and prolong the service life of solar modules and arrays by accurately assessing the health and performance with the PVA-1500 PV analyzer I-V curve tracers. These high-precision analyzers use I-V curve tracing, the most complete solar module performance measurement, to quickly obtain detailed performance data of PV modules. Use the I-V curve tracers to pinpoint problems early to maximize solar energy production while minimizing downtime.

Clamp the included wireless SolSensor™ PV reference sensor to the module frame to measure irradiance and module temperature. With the angular response of each unit calibrated for rotation and elevation, the temperature-compensated sensor offers excellent accuracy over a broad range of technologies, sky conditions, and sun angles.

A portable design and an intuitive interface make it easy to use the analyzer in the field. The PV analyzer and the SolSensor sensor communicate wirelessly with a tablet or a laptop (Windows® only) via Wi-Fi. Download the Data Analysis Tool software to display the data in multiple, easy-to-read formats and a statistical overview of the entire array. The software looks at all the collected data for comparison to see where there are differences, identifying the issue for further troubleshooting. Easily create and send customized reports.

The high-efficiency PV analyzer is ideal for testing high-efficiency modules (>19% module efficiency). The high-efficiency modules have a large in-rush current when you start the test that can overheat the test instrument and slow down testing. Uniquely designed high-efficiency PV analyzer measures all string types, including high-efficiency modules, up to 30 A.

Purchase a high-efficiency PV analyzer I-V tracer with three years of premium care to help reduce unplanned expenses and unexpected downtime. With premium care you get hassle-free, no-charge repair services including labor, parts, and priority shipping. Not only does premium care repair the instrument, one calibration and performance check are also included free of charge and the accessories that originally shipped with the unit will be replaced if they are found to be defective by the technicians.

Premium Care includes:
• Repair manufacturing defects • Annual calibration or performance test • Accident repair • Expedited calibration and repair • Expedited freight • Replacement of damaged accessories • Software updates • Priority tech support

*With module efficiency of <19% for PV analyzer 17785-03.

Conformite Europeenne1 Year Warranty
Specifications
  • DC Min (Volts)20
  • DC Max (Volts)1500
  • DC Voltage Accuracy±(0.5% + 0.25 V)
  • DC Max (amps)30
  • DC Current Accuracy±(0.5% + 0.04 A)
  • InterfaceWi-Fi
  • Safety RatingCAT III 1500 V; IEC 61010-1: Pollution Degree 2
  • PowerRechargeable battery
  • Width (in)8.5
  • Height (in)21
  • Depth (in)6
  • Width (cm)21.6
  • Height (cm)53.3
  • Depth (cm)15.2
  • DescriptionSolmetric PVA-1500/FPC3 High-Efficiency PV Analyzer I-V Curve Tracer with 3 Yr Premium Care
  • Warranty1 year standard + 3 years Premium Care
Reviews
Q&A