Fluke Calibration Portable Calibration Baths
  • The precision and stability of a liquid bath in an affordable, portable package!
These calibration baths are designed with the stability and accuracy of a fluid bath and the compact size and weight of a portable calibrator. Smaller and lighter than most dry-well calibrators, these calibration baths are easy to carry and are spill-proof. The heating, cooling, and stabilization times allow you to take readings within 45 minutes.
Bath is ideal for glass and bimetal thermometers. There’s no need to make custom well inserts — the 2-½" diameter stirred tank holds just about any size probe.
All calibration baths have built-in RS-232 interface and include software for Windows® 3.x/95. The basic data collection software allows you to change the temperature, set points, and safety cutouts on your bath.
Available for sale in the United States and Canada only.
$11,161.89 - $17,209.49CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° F) Temperature Accuracy Temperature Resolution (° F)
Availability Pricing
Fluke Calibration 6102 Portable Calibration Bath; 35 to 200C 9091200 Mfr # 1656950
COMPARE
392 ± 0.25 °C 0.01 °F
$11,161.89CAD / Each
Fluke Calibration 7103 Portable Calibration Bath; -30 to 125C 9091204 Mfr # 7103
COMPARE
257 ± 0.25 °C 0.01 °F
$17,209.49CAD / Each