Fluke Calibration (formerly Hart Scientific) Field Temperature Dry-Well Calibrators
These new units beat every other comparable dry-well in the industry in performance, size, weight, convenience, ease of calibration, software and price. In addition, the heating and cooling rate of each of these dry-wells is adjustable from the front panel, thermal switches can be checked by these units for actuation testing and multiple-hole inserts are available for a variety of probe sizes.
Choose one of three removable inserts sized from 1/16" to 1/2" diameter. Insert A handles a full range of probe sizes with a single well of each size. Insert B features two wells each of 3/8", 1/4" and 3/16" in diameter for doing comparison calibrations, and insert C has six 1/4" diameter wells for multiple probe calibrations.
FEATURES
  • Accuracy to ± 0.5 °C
  • RS232 and Windows® software included
  • Easy to recalibrate
  • Less than eight pounds
$6,721.61 - $7,726.46CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° F) Temperature Accuracy Temperature Resolution (° F)
Availability Pricing
Fluke Calibration 9103-A Dry-Well Calibrator, -13 to 284F/Insert A RK-16101-51 Mfr # 9103-A-156
COMPARE
284
$7,726.46CAD / Each
Fluke Calibration 9140-A Metrology Dry-Well Calibrator RK-16101-70 Mfr # 9140-A-156
COMPARE
662
$6,721.61CAD / Each