Fluke Calibration 5615 Series - Secondary reference PRT/RTD probes

5615 Series - Secondary reference PRT/RTD probes
These probes are the workhorse of secondary calibration laboratories around the world. With a calibration uncertainty at 0°C of just ±0.012°C, the 5615 is a great reference standard for comparison calibrations in a bath or dry-well. The 5615 comes in your choice of 12, 9, or 6 inch sheath lengths.
The 12-inch and 9-inch probes range from –200 to 420°C, and the 6-inch covers the temperature range from –200 to 300°C. The 12-inch probes have a sheath diameter of 1⁄4", and the 9- and 6-inch probes have a sheath diameter of 3⁄16".

Available for sale in the United States and Canada only.
$2,640.61CAD / Each

8 variations of this product are available.

See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connector Type Max Temperature (° F) Description
Availability Pricing
Fluke Calibration 5615-12-L Platinum RTD 12" Probe 1050868 Mfr # 5615-12-L
COMPARE
Mini Spade Lug 788 Platinum RTD 12" Probe
$2,640.61CAD / Each
Fluke Calibration 5615-9-L High Accuracy Temperature Probe 1050869 Mfr # 5615-9-L
COMPARE
Mini Spade Lug 788 High Accuracy Temperature Probe
$2,640.61CAD / Each
Fluke Calibration 5615-12-A Secondary Reference PRT Probe, 12" In-Con 9045024 Mfr # 5615-12-A
COMPARE
Info-Con 788 Secondary Reference PRT Probe, 12" In-Con
$2,640.61CAD / Each
Fluke Calibration 3002846 PRT/RTD Bath Probe, 0.25" Dia x 12" L; 5-Pin 9045026 Mfr # 5615-12-D
COMPARE
5-Pin DIN 788 PRT/RTD Bath Probe, 0.25" Dia x 12" L; 5-Pin
$2,640.61CAD / Each
Fluke Calibration 3376898 PRT/RTD Bath Probe, 0.25" Dia x 12" L; I-Con 9045028 Mfr # 5615-12-P.
COMPARE
Info-Con 788 PRT/RTD Bath Probe, 0.25" Dia x 12" L; I-Con
$2,640.61CAD / Each
Fluke Calibration 3002760 Secondary Reference PRT Probe, 9" , 5 Pin Din 9045035 Mfr # 5615-9-D
COMPARE
5-Pin DIN 788 Secondary Reference PRT Probe, 9" , 5 Pin Din
$2,640.61CAD / Each
Fluke Calibration 3376914 Secondary Reference PRT Probe, 9" Info-Con 9045037 Mfr # 5615-9-P
COMPARE
Info-Con 788 Secondary Reference PRT Probe, 9" Info-Con
$2,640.61CAD / Each
Fluke Calibration 5615-6-D High-Precision Platinum RTD 6" Probe 9320117 Mfr # 5615-6-D
COMPARE
5-Pin DIN 788 High-Precision Platinum RTD 6" Probe
$2,640.61CAD / Each
Showing 1- 8 of 8

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat