Fluke Calibration 3002846 PRT/RTD Bath Probe, 0.25" Dia x 12" L; 5-Pin

Representative image only.

Fluke Calibration 3002846 PRT/RTD Bath Probe, 0.25" Dia x 12" L; 5-Pin

Fluke Calibration | Mfr: 5615-12-D| Item: 9045026

$2,640.61CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$2,640.61CAD / Each
Product Description

Key Features

  • 1 year warranty

More About this Item

5615 Series - Secondary reference PRT/RTD probes
These probes are the workhorse of secondary calibration laboratories around the world. With a calibration uncertainty at 0°C of just ±0.012°C, the 5615 is a great reference standard for comparison calibrations in a bath or dry-well. The 5615 comes in your choice of 12, 9, or 6 inch sheath lengths.
The 12-inch and 9-inch probes range from –200 to 420°C, and the 6-inch covers the temperature range from –200 to 300°C. The 12-inch probes have a sheath diameter of 1⁄4", and the 9- and 6-inch probes have a sheath diameter of 3⁄16".

Available for sale in the United States and Canada only.

Hart Scientific is now Fluke Calibration.

1 Year Warranty
Specifications
  • Probe TypePRT/RTD
  • Min Temperature (° F)-328
  • Max Temperature (° F)788
  • Min Temperature (° C)-200
  • Max Temperature (° C)420
  • Temperature Accuracy±0.022°F (±0.012°C)
  • Connector Type5-Pin DIN
  • Sheath Diameter (InchShort)0.25
  • DescriptionPRT/RTD Bath Probe, 0.25" Dia x 12" L; 5-Pin
  • Warranty1 year
Reviews
Q&A