Fluke Calibration 5606 Series - Full immersion PRT/RTD probes
5606 Series - Full immersion PRT/RTD probes: Designed to perform in extreme environments where both the transition junction and the lead wires are required to withstand temperatures covering the entire operating range of the probe. Applications include calibration or validation of sensors used in laboratory or bio freezers, walk-in refrigerators, autoclaves, ovens, stability test chambers, furnaces and incubators.
Available for sale in the United States and Canada only.
$1,149.24CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connector Type Max Temperature (° F)
Availability Pricing
Fluke Calibration 3622830 PRT/RTD Freezer Probe, 0.125" Dia x 2" L; Wire 9045000 Mfr # 5606-50-B
COMPARE
Stripped Wire 320
$1,149.24CAD / Each
Fluke Calibration 3622907 PRT/RTD Probe, 0.125" D x 2" L 9045004 Mfr # 5606-50-P
COMPARE
Info-Con 320
$1,149.24CAD / Each