Fluke 77-IV General Purpose Digital Multimeter
Available for sale in the United States and Canada only Versatile meter for field service or bench repair
  • Wide 1000 V measurement range
  • Average responding AC measurements
  • 0.3% accuracy
  • 10 Amps continuous (20A for 30 seconds)
  • Frequency and capacitance
  • Resistance and continuity
  • Large display backlight for work in dim areas
  • Min / Max to record signal fluctuations
  • Ergonomic over molded housing with integrated probe holders
  • Auto and Manual ranging
  • Meets measurement category 1000 V CAT III and category IV 600 V safety standards
The Fluke 77-IV digital multimeter has the features needed to repair most electrical and electronic problems. This meter is simple to use and features more measurement functions, conformance to the latest safety standards, and a large display that's easy to view.
Available for sale in the United States and Canada only.
$668.92CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
DC Max (Volts) DC Voltage Accuracy True Rms
Availability Pricing
Fluke 77-4 General Purpose Digital Multimeter with Backlight 2000571 Mfr # FLUKE-77-4
COMPARE
1000 ±(0.3% + 1)
$668.92CAD / Each