Fluke 719Pro Electric Pressure Calibrators
Unique integrated electric pump for one-handed pressure calibration to 300 psi
  • Easy-to-use switch test function for checking pressure switches
  • Precision pressure adjust vernier for easy and accurate pressure calibration
  • Obtains ±0.025% accuracy for reference class transmitter calibration
  • Covers 30 psi, 150 psi and 300 psi pressure ranges
  • Pressure measurement ranges are extended when used with any of 29 700Pxx pressure modules
  • Pump cleanout ports and fluid trap reduce pump failures from moisture and fluid damage
The Fluke® 719Pro electric pressure calibrator is the ideal test tool for calibrating high-accuracy transmitters, pressure switches and pressure gauges. This instrument includes a full-functioning loop calibrator that sources, simulates and measures mA signals and more.
Large backlit screen displays three parameters at once: pressure measurement from an internal or external sensor, sourced/simulated or measured mA values, and temperature measured by an optional RTD probe.
3 PRODUCT OPTIONS
Compareitem specifications
Max Pressure Pressure Accuracy Pressure Resolution
Availability Pricing
Fluke 719PRO 30G Electrical Pressure Calibrator, 0 to 30 psig 1610715 Mfr # Fluke-719PRO 30G
COMPARE
30 psi (2.0 bar) ±0.025 % of fullscale, 6 months±0.035 % of fullscale, 1 year 0.001 psi (0.0001 bar)
$8,190.43CAD / Each
Fluke 719PRO 150G Electrical Pressure Calibrator, 0 to 150 psig 1610716 Mfr # Fluke-719PRO 150G
COMPARE
150 psi (10 bar) ±0.025 % of fullscale, 6 months±0.035 % of fullscale, 1 year 0.01 psi (0.001 bar)
$8,190.43CAD / Each
Fluke 719PRO 300G Electrical Pressure Calibrator, 0 to 300 psig 1610721 Mfr # Fluke-719PRO 300G
COMPARE
300 psi (20 bar) ±0.025 % of fullscale, 6 months±0.035 % of fullscale, 1 year 0.01 psi (0.001 bar)
$8,190.43CAD / Each