Fluke® 718 Series Pressure Calibrators
Best in class mA accuracy (0.015%) for better measurement confidence and workload coverage
  • Pressure source and milliamp measurement capability that calibrates and maintains almost any pressure device│Integrated pump with media protection│Error % calculation for quicker pass/fail decision making
The Fluke® 718 Pressure Calibrator provides a total pressure calibration solution for transmitters, gauges and switches. This compact pressure calibration solution is about a third of the size of comparable instruments and weighs just one kilogram (two pounds). The unique easy to clean pump helps protect the pump from damage and provides the ability to service the pump without disassembly, even in the field.
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Measurement (mA) Max Pressure Min Pressure
Availability Pricing
Fluke 718 100US Pressure Calibrator, 100 psig 6845087 Mfr # FLUKE-718 100US
COMPARE
24 100 psig -12
$5,759.48CAD / Each
Fluke 718 30US Pressure Calibrator, 30 psig 6845089 Mfr # FLUKE-718 30US
COMPARE
24 30 psig -12
$5,759.48CAD / Each
Fluke 718 300G Pressure Calibrator, 300 psig 6845090 Mfr # FLUKE-718 300G
COMPARE
24 300 psig -12
$5,759.48CAD / Each