Fluke 718 30US Pressure Calibrator, 30 psig
Representative image only.
Fluke 718 30US Pressure Calibrator, 30 psig
Fluke | Mfr: FLUKE-718 30US| Item: 6845089
Best in class mA accuracy (0.015%) for better measurement confidence and workload coverage
$5,932.25CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$5,932.25CAD / Each
Buy It Together
Current Item
Fluke 718 30US Pressure Calibrator, 30 psig
Fluke 718 30US Pressure Calibrator, 30 psig
6845089
$5,932.25CAD / Each
+ =
Total Price 6288.81 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Pressure source and milliamp measurement capability that calibrates and maintains almost any pressure device│Integrated pump with media protection│Error % calculation for quicker pass/fail decision making
  • 3 year warranty
More About this Item
The Fluke® 718 Pressure Calibrator provides a total pressure calibration solution for transmitters, gauges and switches. This compact pressure calibration solution is about a third of the size of comparable instruments and weighs just one kilogram (two pounds). The unique easy to clean pump helps protect the pump from damage and provides the ability to service the pump without disassembly, even in the field.
3 Year Warranty
Specifications
  • Max Measurement (mA)24
  • Max Pressure30 psig
  • Min Pressure-12
  • Display Type5-digit LCD
  • BatteriesTwo 9V alkaline
  • Battery life4 to 20 hours
  • DescriptionPressure Calibrator, 30 psig
  • Warranty3 Years