Fluke 62 MAX Infrared Thermometers
Small and lightweight, yet extremely accurate and very easy to use
  • Dust and water-resistant
  • Drop tested to 3 meters (9.8-foot)
  • Small and lightweight
Temperature is often the first indication of a potential problem. The Fluke 62 MAX noncontact thermometers are ideal for quick, basic temperature checks in applications where the user needs to be a safe distance from the target area being measured. This compact, portable tool enables professionals to scan surfaces for hot spots, diagnose problems, and monitor temperature variations without direct contact. Easy to use—just aim the laser sighting at the center of the target area, pull the trigger, and read the temperature on the large backlit LCD.
IP54 rated for dust and water resistance, ruggedly built to withstand a 3-meter drop and ergonomic designed for a more natural hand fit makes the Fluke 62 MAX meters ideal for use in many different work environments. These IR thermometers feature Fahrenheit or Celsius selectable units, current/min/max temperature readings, average and differential temperatures, adjustable high/low setpoints with audible alarm, seven-second auto hold, continuous scanning, and power off.
$225.41CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Distance To Target Size Ratio (D:S) Emissivity Max Temperature (° F)
Availability Pricing
Fluke 62 MAX Compact Infrared Thermometer (10: 1) 3964116 Mfr # FLUKE-62 MAX
COMPARE
10:1 Adjustable 932
$225.41CAD / Each
Fluke 62 MAX+ Compact Infrared Thermometer (12: 1) 3964117 Mfr # FLUKE-62 MAX+
COMPARE
12:1 Adjustable 1202