Fluke 381 Clamp Meter
The most innovate, versatile, diagnostic tool with increased performance and flexibility
Fluke 381 Clamp Meter is the first clamp meter with a detachable, remote display for easier, faster, safer measurements. Meter allows you to conduct two jobs simultaneously; clamp the meter around a conductor, remove the display, and walk across the room to operate controls or remove protective equipment, all while watching real-time readings. Includes new iFLEX flexible current probe expanding the measurement range to 2500 A ac while providing increased flexibility to measure awkward sized conductors.
Fluke iFLEX Flexible Current Probes expand the measurement range of select Fluke clamp meters to 2500 A ac providing additional flexibility. iFLEX probes connect directly to the clamp meter, displaying current measurements without error prone scaling factors. Features large loop size and thin cross section for awkward sized conductors and improved access in crowded spaces.
Available for sale in the United States and Canada only.
$1,337.84CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
DC Max (Volts) DC Voltage Accuracy True Rms
Availability Pricing
Fluke 381 True-RMS 1000A AC/DC Clamp Meter with iFlex 2500 Probe 2003705 Mfr # FLUKE-381
COMPARE
1000
$1,337.84CAD / Each