Fluke 280 Series True-RMS Digital Logging Multimeters
Industrial Meter for demanding users
  • Built-in logger records reading and time of day to catch intermittent problems
  • View data on the meter in TrendCapture or with optional FlukeView software
  • Features peak capture to measure transients as fast as 250 µS
  • Maximize productivity with the convenient Fluke 289/FVF combination kit
The 280 Series multimeters are designed for the changing needs of today’s professionals involved in the solving complex problems in electronics, plant automation, power distribution, and manufacture and design of electronic circuits or systems. These True-RMS 50,000-count multimeters feature a large, bright white display with dual parameter readout. Use TrendCapture function for characterizing the performance of a design, or for unattended monitoring while in the field, and to graphically view logged readings without a PC. Transfer data to your PC with optional FlukeView forms software for further analysis.
Available for sale in the United States and Canada only.
Additional features include internal memory that logs up to 200 hours, 0.025% DC voltage accuracy with 1 microvolt resolution, and “i” button for instant access to on-board help screens.
Model 289 also features low pass filter, LoZ function for false readings due to ghost voltage, and 50 Ω range for accurate motor winding measurements.
Available for sale in the United States and Canada only
$1,488.11 - $1,797.52CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
AC Max (Volts) DC Max (Volts) True Rms
Availability Pricing
Fluke 289/IMSK Digital Multimeter Service Kit with Current Clamp 2003437 Mfr # FLUKE-289/IMSK
COMPARE
1000 1000 Yes
$1,576.51CAD / Each
Fluke 287/FVF True-RMS Electronic Logging Digital Multimeter Combo Kit with TrendCapture 2604892 Mfr # FLUKE-287/FVF
COMPARE
1000 1000 Yes
$1,706.17CAD / Each
Fluke 287 True RMS Electronic Logging Digital Multimeter with TrendCapture 2604895 Mfr # FLUKE-287
COMPARE
1000 1000 Yes
Call For Price
Fluke 289 True RMS Industrial Logging Digital Multimeter with TrendCapture 2604896 Mfr # FLUKE-289
COMPARE
1000 1000 Yes
$1,488.11CAD / Each
Fluke 289/FVF True-RMS Industial Logging Digital Multimeter Combo Kit with TrendCapture 2604897 Mfr # FLUKE-289/FVF
COMPARE
1000 1000 Yes
$1,797.52CAD / Each