Fluke 170-Series Digital Multimeters and Kits
General purpose multimeters provide precision, safety & reliability High performance at an affordable price Versatile meter for field service or bench repair
  • Wide 1000 V measurement range
  • Manual or automatic ranging
  • Min / Max to record signal fluctuations
  • Large display - backlight for work in dim areas
  • Ergonomic overmolded housing with integrated probe holders
  • Free your hands with the optional TPAK magnetic hanger
  • Meets Measurement Category 1000 V CAT III and Category IV 600 V safety standards
These multimeters all feature a 6000-count resolution, display and auto hold, frequency, capacitance, resistance, continuity, and diode measurement capability. Meters include easy battery exchange, closed case calibration through the front panel, and an ergonomic case with integrated protective holster.
Models 177 and 179 feature True RMS voltage and current measurement capabilities, and provide digital display with analog bar graph and backlight. Model 77 IV also includes average responding AC measurements, and model 179 features temperature measurement capabilities.
Available for sale in the United States and Canada only.
$668.90 - $804.46CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
DC Max (Volts) DC Voltage Accuracy True Rms
Availability Pricing
Fluke 177 ESFP Digital True RMS Multimeter with Backlight 2000567 Mfr # FLUKE-177 ESFP
COMPARE
1000 ±0.09% True Rms
$721.95CAD / Each
Fluke 175 ESFP True-RMS Digital Multimeter, CAT IV 600V/CAT III 1000V 2607501 Mfr # FLUKE-175 ESFP
COMPARE
1000 ±0.15% True Rms
$668.90CAD / Each
Fluke 179 True RMS Multimeter with backlight and temperature 2607901 Mfr # FLUKE-179 ESFP
COMPARE
1000 ±0.09% True Rms
$774.25CAD / Each
Fluke 179/EDA2 Industrial Electronics Combo Kit 2607904 Mfr # FLUKE-179/EDA2
COMPARE
1000 ±0.09% True Rms
$804.46CAD / Each