Fluke 117-Series Digital Multimeters
True-RMS and a 6000-count display in a compact meter
These compact meters feature True-RMS AC and DC voltage, 6000-count resolution, min/max/average capabilities, frequency, and capacitance. Units include a large display and white LED backlight for working in poorly lit environments.
Fluke Model 117 is ideal for electricians and use in demanding conditions. Unit includes VoltAlert™ technology for integrated noncontact voltage selection, AutoVolt feature provides automatic AC/DC voltage selection. Low input impedance prevents false readings due to ghost voltage.
Available for sale in the United States and Canada only.
1 PRODUCT OPTIONS
Compareitem specifications
DC Max (Volts) DC Voltage Accuracy True Rms
Availability Pricing
Fluke 117 Electrician's Multimeter with True-RMS 2601632 Mfr # FLUKE-117
COMPARE
600 0.5% + 2
$449.38CAD / Each