FLIR® Ex Pro Series Infrared Cameras with Ingnite™ Cloud Connectivity
A perfect tool for locating and diagnosing electrical, mechanical, and building problems
  • MSX®-enhanced thermal images provide amazing detail to help you identify problem areas
  • A 640 x 480 resolution color touchscreen
  • Save time on manual temperature adjustment with one-touch level/span feature
  • Built-in NFPA 70B compliant ?T measurement tools
  • Bright 9 cm (3.5”) LCD for easy viewing
  • IP54 rated for protection against dust and water
  • Designed to withstand a 2-m (6.5-ft) drop
  • Upload files to FLIR Ignite cloud over Wi-Fi
  • Battery lasts up to 4 hours
Quickly pinpoint hot spots and easily identify problems with the E5 Pro and E8 Pro thermal imaging cameras. Use these rugged, handheld IR cameras for monitoring and preventive maintenance in building surveys, HVAC, automotive, and general electrical and mechanical inspections. Select from the following modes to view your images: thermal MSX (Multi Spectral Dynamic Imaging), thermal only, picture-in-picture, or digital camera. In the MSX mode, the edges of the objects are enhanced with visual image details, providing more crisp images. Use the picture-in-picture mode to view an infrared image on top of the visual image.
The thermal imagers have features to simplify measurements and help save time. The one-touch level/span feature allows you to choose a small area of focus in a thermal image, then with a single touch of the screen, the camera will auto-adjust the level, saving you time on manual adjustment. Calculate ?T on the camera without any additional steps using the built-in NFPA 70B compliant Delta T measurement tools.
The 9 cm (3.5”), 640 x 480 resolution color touchscreen LCD is easy to use and helps improve inspection efficiency. Add detailed notes, edit and organize photos, and upload files directly from the camera to the FLIR Ignite cloud over Wi-Fi for convenient access from any device. Share images and create quick reports in FLIR Ignite or use FLIR Thermal Studio software to create professional thermal images and reports from a PC.
$1,987.60CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Pixels / Thermal Resolution Thermal Sensitivity (N.E.T.D) (° C)
Availability Pricing
Flir FLIR E5 Pro Infrared Camera with Ignite™ Cloud Connection, 160 x 120 IR Resolution 3975932 Mfr # 13301-0101
COMPARE
400 160 x 120 (19,200 pixels) <0.06 °C / <60 mK at 30 °C
$1,987.60CAD / Each