Extech Waterproof Temperature Indicators
Won't fail under occasional splashing
  • Waterproof, dustproof housing is IP65-rated
  • Won't permit out-of-range failures—visual and audible alarms alert you to under- and over-range conditions
  • Goes where you need it—magnetic back for hanging, built-in stand for benchtop, or hang directly on wall
  • Choose NSF-certified model 39758-99 for use in the food service industry
Temperature indicator features selectable temperature scale (°F/°C), high/low set points, min/max reading, data hold, alarm icons, and low-battery indicator.
$44.20 - $61.88CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Min Temperature (° F) Description
Availability Pricing
Extech TM26 NSF Certified Waterproof Temperature Indicator 3975899 Mfr # TM26
COMPARE
392 -40 NSF Certified Waterproof Temperature Indicator
$61.88CAD / Each
Extech TM20 Waterproof Thermometer; 3/4" SS Probe, 9.2' Cable 9500155 Mfr # TM20
COMPARE
158 -40 Waterproof Thermometer; 3/4" SS Probe, 9.2' Cable
$44.20CAD / Each