Extech RF EMF Strength Meter
High Frequency measurement for EMF
  • For electromagnetic field strength measurement including mobile phones, cell phones, base stations, and microwave leakage
  • Non-directional (isotropic) measurement with three-channel (triaxial) measurement probe
  • Max Hold and Average functions
  • Manual store/recall up to 99 sets
  • Level exceed audible alarm with user selectable threshold
  • Includes 9V battery and carrying case
  • Applications
  • RF electromagnetic wave field strength measurements
  • Mobil phone base station antenna radiation power density measurement
  • Radiation safety level tests
  • Locating RF ''Hot Spots'' in the workplace or at home
  • Microwave oven leakage tests
  • RF ''Electrosmog'' detection
  • Wireless network (Wi-Fi) RF detection
$514.21 - $1,206.71CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Extech 480836 RF EMF Strength Meter 9.3" L x 2.4" W x 2.4" H (237 x 60 x 60mm) 2682240 Mfr # 480836
COMPARE
RF EMF Strength Meter 9.3" L x 2.4" W x 2.4" H (237 x 60 x 60mm)
$514.21CAD / Each
Extech 480846 RF EMF Strength Meter, 8 GHz 9.3" L x 2.4" W x 2.4" H (237 x 60 x 60mm) 2682242 Mfr # 480846
COMPARE
RF EMF Strength Meter, 8 GHz 9.3" L x 2.4" W x 2.4" H (237 x 60 x 60mm)
$1,206.71CAD / Each