Extech Mini Thermoanemometers
Rugged pocket-size meters fold for convenient storage
  • Water- and shock-resistant housing provides excellent durability
  • Fold-up case acts as an extension handle and protects the meter during storage
  • Large dual display of air velocity and temperature
  • Data Hold and Auto Power-off
  • Water-resistant housing floats and drop-tested to 6 feet
  • Windspeed: Measures ft/min, MPH, m/s, km/h, knots and Beaufort Force
  • Windspeed: Selectable averaging function of 5, 10, or 13 second intervals
  • Winspeed: Replaceable non-corrosive plastic wind vane (impeller)
  • Measures temperature and windchill from 0 to 122°F (–18 to 50°C)
These anemometers feature a dual LCD to display air velocity with temperature, and windchill. Feature selectable average function, one-second display update, maximum reading, hold function, and auto shutoff after 20 minutes.
$204.80 - $293.20CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Air Velocity Accuracy Air Velocity Resolution Max Air Velocity (ft/min)
Availability Pricing
Extech 45118 Mini Thermoanemometer with Temperature 1020110 Mfr # 45118
COMPARE
±(3% of reading + 40 ft/min or 0.2 m/s) 20 ft/min (0.01 m/s, 0.2 mph, 0.7 km/h, 0.3 knots, 1 BF) 5500
$204.80CAD / Each
Extech 45158 Mini Thermoanemometer with Temperature and Humidity 1020120 Mfr # 45158
COMPARE
±(3% of reading + 40 ft/min or 0.2 m/s) 20 ft/min (0.01 m/s, 0.2 mph, 0.7 km/h, 0.3 knots, 1 BF) 5500
$293.20CAD / Each