Environmental Express® Katanax® Platinumware Polishing Box Kits
Restore your platinumware to its original shine
  • Versatile polishing for any platinum-group metals, not just fusion platinumware
  • Designed for semi-permanent installation on your workbench, providing stability and efficiency
Elevate the maintenance of your laboratory equipment with the Katanax® Polishing Kit – an ultimate solution for restoring the bright finish on platinum crucibles, molds, and any items made from platinum-group metals. This versatile kit is designed to handle all brands and shapes, making it an essential tool for any lab. The kits include a powerful rotary electric tool with multiple attachments and a specialized polishing station that rotates your platinumware, ensuring an even, brilliant finish every time.
$3,735.04 - $4,442.27CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Compatible With Description
Availability Pricing
Environmental Express Katanax® Platinumware Polishing Box Kit, 115V 0446856 Mfr # KP9031A
COMPARE
Platinum crucibles and molds Platinumware Polishing Box Kit, 115V
$3,735.04CAD / Each
Environmental Express Katanax® Platinumware Polishing Box Kit, 230V 0446857 Mfr # KP9032A
COMPARE
Platinum crucibles and molds Platinumware Polishing Box Kit, 230V
$3,911.85CAD / Each
Environmental Express Katanax® Platinumware Polishing Box Kit, 220V 0446858 Mfr # KP9033A
COMPARE
Platinum crucibles and molds Platinumware Polishing Box Kit, 220V
$4,442.27CAD / Each
Environmental Express Katanax® Platinumware Polishing Box Kit without Rotary Tool 0446859 Mfr # KP9034A
COMPARE
Platinum crucibles and molds Platinumware Polishing Box Kit without Rotary Tool
$4,059.19CAD / Each