Environmental Express Katanax® Platinumware Polishing Box Kit, 220V

Representative image only.

Environmental Express Katanax® Platinumware Polishing Box Kit, 220V

Environmental Express | Mfr: KP9033A| Item: 0446858

Restore your platinumware to its original shine

$4,442.27CAD / Each

$4,442.27CAD / Each
Product Description

Includes

Rotary tool, flex shaft extension, vertical stand, brush pads, diamond pastes, felt pads, microfiber clothes, mineral oil, polishing box, mold size fixtures, crucible fixture, and pin vices handle.

Key Features

  • Versatile polishing for any platinum-group metals, not just fusion platinumware
  • Designed for semi-permanent installation on your workbench, providing stability and efficiency

More About this Item

Elevate the maintenance of your laboratory equipment with the Katanax® Polishing Kit – an ultimate solution for restoring the bright finish on platinum crucibles, molds, and any items made from platinum-group metals. This versatile kit is designed to handle all brands and shapes, making it an essential tool for any lab. The kits include a powerful rotary electric tool with multiple attachments and a specialized polishing station that rotates your platinumware, ensuring an even, brilliant finish every time.

Conformite Europeenne
Specifications
  • Compatible WithPlatinum crucibles and molds
  • Power (VAC)220
  • DescriptionPlatinumware Polishing Box Kit, 220V
Reviews
Q&A