Environmental Express High-Performance Combination pH Electrodes
Affordable, high-accuracy pH electrodes for every application
  • The ideal 5-point calibration pH electrode
  • Delivers superior reproducibility in aqueous samples
Environmental Express high-performance electrodes are available in a full range of materials, junctions, and sizes to suit your application requirements. New formulation provides additional stability and superior reproducibility over entire pH range. Fill solution purchased seperately, 05992-47 for single junction, 05992-48 for double juncion electrodes
$268.82 - $767.78CAD / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Electrode Body Material Electrode Type Description
Availability Pricing
COMPARE
Glass Single junction pH Electrode,12x110mm, Glass Body, Single Junction, Refill
$373.65CAD / Each
COMPARE
Epoxy Single junction pH Electrode,12x110mm, Epoxy Body, Single Junction, Refill
$343.89CAD / Each
COMPARE
Epoxy Double junction pH Electrode,12x110mm, Epoxy Body, Double Junction, Refill
$451.37CAD / Each
COMPARE
Epoxy Single junction pH Electrode,12x110mm, Epoxy Body, Single Junction, Sealed
$275.38CAD / Each
COMPARE
Epoxy Single junction pH Electrode,6x110mm, Epoxy Body, Single Junction, Sealed
$353.54CAD / Each
COMPARE
Epoxy Double junction pH Electrode,12x110mm, Epoxy Body, Double Junction, Sealed
$268.82CAD / Each
COMPARE
Glass Double junction Fast-response pH Electrode,12x110mm, Glass Body, Double Junction, Refill
$653.30CAD / Each
COMPARE
Glass Double junction Fast-response pH Electrode,6x110mm, Glass Body, Double Junction, REfill
$767.78CAD / Each
COMPARE
Epoxy Double junction Fast-response pH Electrode,12x110mm, Epoxy Body, Double Junction, Sealed
$598.71CAD / Each
COMPARE
Glass Single junction Rugged pH Electrode,12x110mm, Glass Body, Single Junction, Refill
$398.40CAD / Each
COMPARE
Glass Double junction Rugged pH Electrode,12x110mm, Glass Body, Double Junction, Refill
$420.06CAD / Each
Showing 1- 11 of 11