Environmental Express pH Electrode,6x110mm, Epoxy Body, Single Junction, Sealed
Representative image only.
Environmental Express pH Electrode,6x110mm, Epoxy Body, Single Junction, Sealed
Environmental Express | Item: 5551022
Affordable, high-accuracy pH electrodes for every application
$353.54CAD / Each
$353.54CAD / Each
Buy It Together
Current Item
Environmental Express pH Electrode,6x110mm, Epoxy Body, Single Junction, Sealed
Environmental Express pH Electrode,6x110mm, Epoxy Body, Single Junction, Sealed
5551022
$353.54CAD / Each
+ =
Total Price 451.52 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Includes
3.3-ft (1-m) cable with a BNC connector.
Key Features
  • The ideal 5-point calibration pH electrode
  • Delivers superior reproducibility in aqueous samples
  • 1 year warranty
More About this Item
Environmental Express high-performance electrodes are available in a full range of materials, junctions, and sizes to suit your application requirements. New formulation provides additional stability and superior reproducibility over entire pH range. Fill solution purchased seperately, 05992-47 for single junction, 05992-48 for double juncion electrodes
ISO9001 Certified Supplier1 Year Warranty
Specifications
  • ApplicationsSealed
  • Electrode TypeSingle junction
  • Electrode Body MaterialEpoxy
  • Min pH (pH)0
  • Max pH (pH)14
  • Min Temperature (° C)0
  • Max Temperature (° C)80
  • ConnectorBNC
  • Diameter (mm)6
  • Length (mm)110
  • Cable Length (m)1
  • DescriptionpH Electrode,6x110mm, Epoxy Body, Single Junction, Sealed