Electro-Tech Systems 5504 Series Temperature and Humidity Glove Box Chambers
Ideal for small applications with critical environmental requirements
  • Large front door offers easy access
  • Thick clear acrylic offers excellent visibility to contents
These compact benchtop chamber glove boxes provide a usable work space of 3.25 cubic feet (92 L). Select a model to meet your humidity and temperature performance needs—use as a basic glove box or as a complete environmental chamber with humidity and temperature controls.
The large 18”W x 12”H (46 x 30 cm) access door features two 6″ (15 cm) ports which can be configured for gloves or iris ports. Interior work space is 23.5”W x 14.5”H x 17.25”D (60 x 37 x 44 cm).
$11,134.41 - $25,893.36CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Min Relative Humidty (%) Power (VAC) Description
Availability Pricing
Electro-Tech Systems 5504-7614 Glove Box Chamber, Humidity (5 to 95% RH) and Temperature (10 to 60°C) Controlled, Self-Regenerating Dehumidification; 115V RK-03322-93 Mfr # 0687437927614
COMPARE
5 115 Glove Box Chamber, Humidity (5 to 95% RH) and Temperature (10 to 60°C) Controlled, Self-Regenerating Dehumidification; 115V
$25,893.36CAD / Each
Electro-Tech Systems 5504-7607 Glove Box Chamber, Humidity (10 to 95% RH) and Temperature (10 to 60°C) Controlled, Desiccant/Pump Dehumidification; 115V RK-03322-94 Mfr # 0687437927607
COMPARE
10 115 Glove Box Chamber, Humidity (10 to 95% RH) and Temperature (10 to 60°C) Controlled, Desiccant/Pump Dehumidification; 115V
$21,190.29CAD / Each
Electro-Tech Systems 5504-7645 Glove Box Chamber, Humidity (10 to 95% RH) Controlled, Desiccant/Pump Dehumidification, No Temperature Control; 115V RK-03322-95 Mfr # 0687437927645
COMPARE
10 115 Glove Box Chamber, Humidity (10 to 95% RH) Controlled, Desiccant/Pump Dehumidification, No Temperature Control; 115V
$13,888.17CAD / Each
Electro-Tech Systems 5504-7652 Glove Box Chamber, Humidity (10% to ambient RH) Controlled, Desiccant/Pump Dehumidification, No Temperature Control; 115V RK-03322-96 Mfr # 0687437927652
COMPARE
10 115 Glove Box Chamber, Humidity (10% to ambient RH) Controlled, Desiccant/Pump Dehumidification, No Temperature Control; 115V
$11,134.41CAD / Each
Electro-Tech Systems 5504-7638 Glove Box Chamber, Humidity (5 to 95% RH) and Temperature (10 to 60°C) Controlled, Self-Regenerating Dehumidification; 230V RK-03322-97 Mfr # 0687437927638
COMPARE
5 230 Glove Box Chamber, Humidity (5 to 95% RH) and Temperature (10 to 60°C) Controlled, Self-Regenerating Dehumidification; 230V
$25,893.36CAD / Each
Electro-Tech Systems 5504-7621 Glove Box Chamber, Humidity (10 to 95% RH) and Temperature (10 to 60°C) Controlled, Desiccant/Pump Dehumidification; 230V RK-03322-98 Mfr # 0687437927621
COMPARE
10 230 Glove Box Chamber, Humidity (10 to 95% RH) and Temperature (10 to 60°C) Controlled, Desiccant/Pump Dehumidification; 230V
$21,190.29CAD / Each