Edwards Vaccum Centering Rings
Create a tight vacuum seal with these centering rings
Polymer support and nitrile O-rings withstand up to 10–7 Torr (mm Hg). The temperature range of these centering rings is 14 to 212°F (–10 to 100°C).
The centering ring has an elastomer O-ring creating the seal, and inside of this there is a plastic support ring.
$26.67 - $143.80CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Fitting Material Connection Type Description
Availability Pricing
Edwards C100-07-173 Centering Ring, 63 ISO-K - Nitrile (Vacuum Pump Accessories) RK-31401-82 Mfr # C100-07-173
COMPARE
Nitrile Vacuum Flange Centering Ring, 63 ISO-K - Nitrile (Vacuum Pump Accessories)
$143.80CAD / Each
Edwards C105-12-393 Centering ring, NW 10/16 - Nitrile (Vacuum Pump Accessories) RK-31402-70 Mfr # C105-12-393
COMPARE
Nitrile Vacuum Flange Centering ring, NW 10/16 - Nitrile (Vacuum Pump Accessories)
$26.67CAD / Each
Edwards C105-14-393 Centering ring, NW 20/25 - Nitrile (Vacuum Pump Accessories) RK-31402-72 Mfr # C105-14-393
COMPARE
Nitrile Vacuum Flange Centering ring, NW 20/25 - Nitrile (Vacuum Pump Accessories)
$28.14CAD / Each