Dwyer Digihelic II Differential Pressure Controllers
  • APPLICATIONS
  • Building automation systems
  • Pharmaceutical / clean room
  • Semiconductor
  • Fume hoods
  • Dust collection
The Digihelic II is an innovative product that offers three instruments in one NEMA 4X Enclosure. The unit combines a digital display, relay outputs, and a current output all in one unit. The series DHII offers a digital upgrade to the industry standard Photohelic differential pressure switch/gage. This product saves space, reduces inventory, and saves installation money with its combination of features. In addition to the 3-in-1 technology the Digihelic II also allows selection of engineering units, has a universal power supply, and a selectable display reading pressure, velocity or flow measurements. Storage of maximum and minimum pressure values is also standard, as well as Modbus serial port communication. Optional software is available for the changing of any programmable setting via a remote computer.
$1,709.13 - $1,989.08CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max Pressure (PSI) Resolution
Availability Pricing
Dwyer DHII-006 Digihelic Differential Pressure Controller, 5" WC 6843712 Mfr # DHII-006
COMPARE
±0.5% full-scale
$1,989.08CAD / Each
Dwyer DHII-007 Digihelic Differential Pressure Controller, 10" WC 6843713 Mfr # DHII-007
COMPARE
±0.5% full-scale
$1,709.13CAD / Each