Diversey Soft Care® Foam Instant Hand Sanitizer

Just rub in and dry—no water or rinsing needed
  • Contains alcohol for fast drying
  • Moisturizer helps reduce skin dryness
Soft Care foam hand sanitizer is ideal for use in public areas, schools, sporting events, and food service areas. Simply pump the hand sanitizer onto hands and rub to cover all surfaces. Fast drying sanitizer requires no rinsing or towels. Sanitizer does not contain any added dyes or fragrances to irritate your skin.
$202.59CAD / Case of 6

1 variations of this product are available.

See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Applications Dilution Format
Availability Pricing
Diversey Soft Care® Foam Instant Hand Sanitizer, 18 oz Bottle; 6/CS 7890162 Mfr # 100930835
COMPARE
For use in public areas, schools, sporting events, and food service areas Ready to use Clear, liquid (foam)
$202.59CAD / Case of 6
Showing 1- 1 of 1

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat