Diversey Soft Care® Foam Instant Hand Sanitizer, 18 oz Bottle; 6/CS

Representative image only.

Diversey Soft Care® Foam Instant Hand Sanitizer, 18 oz Bottle; 6/CS

Diversey | Mfr: 100930835| Item: 7890162

Just rub in and dry—no water or rinsing needed

$202.59CAD / Case of 6

$202.59CAD / Case of 6
Product Description

Key Features

  • Contains alcohol for fast drying
  • Moisturizer helps reduce skin dryness
  • pH of 5

More About this Item

Soft Care foam hand sanitizer is ideal for use in public areas, schools, sporting events, and food service areas. Simply pump the hand sanitizer onto hands and rub to cover all surfaces. Fast drying sanitizer requires no rinsing or towels. Sanitizer does not contain any added dyes or fragrances to irritate your skin.

Available for sale to the 48 continental United States only.

Specifications
  • FormatClear, liquid (foam)
  • Volume (Liters)0.53
  • DilutionReady to use
  • ApplicationsFor use in public areas, schools, sporting events, and food service areas
  • DescriptionSoft Care® Foam Instant Hand Sanitizer, 18 oz Bottle; 6/CS
Reviews
Q&A